Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011259.1 | NP_035389.1 | 6 | 767 | ||
GenBank mRNA | AK008050.1 | - | 3 | 422 | ||
AK008140.1 | - | 3 | 422 | |||
AK008470.1 | - | 4 | 602 | |||
D63356.1 | - | 6 | 767 | |||
D63357.1 | - | 6 | 716 |