Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011274.5 | NP_035404.4 | 6, 7 | 851 | ||
GenBank mRNA | AF091096.1 | - | 4, 5 | 373 | ||
AK133383.1 | - | 6, 7 | 526 | |||
AK155994.1 | - | 6, 7 | 825 | |||
AK166566.1 | - | 6, 7 | 842 | |||
AK170929.1 | - | 6, 7 | 846 | |||
BC023029.1 | - | 6, 7 | 511 | 5374731 |