Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB112924.1 | - | 20, 21 | 2074 | ||
AK139040.1 | - | 19, 20 | 2018 | |||
BC042585.1 | - | 20, 21 | 2241 | 5364086 | ||
BC050883.1 | - | 20, 21 | 2208 | 5698459 | ||
D29965.1 | - | 20, 21 | 2051 |