Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011304.3 | NP_035434.1 | 6 | 608 | ||
GenBank mRNA | AB013912.1 | - | 6 | 439 | ||
AK135404.1 | - | 6 | 422 | |||
AK144156.1 | - | 6 | 430 | |||
AK159112.1 | - | 6 | 610 | |||
BC072585.1 | - | 1107 | ||||
BC115809.1 | - | 6 | 426 | 40105495 | ||
BC116693.1 | - | 6 | 425 | 40105493 |