Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081459.2 | NP_001074928.1 | 10 | 3288 | ||
NM_001289538.1 | NP_001276467.1 | 9 | 3188 | |||
NM_001289539.1 | NP_001276468.1 | 10 | 2911 | |||
NM_001289540.1 | NP_001276469.1 | 11 | 3048 | |||
NM_001289541.1 | NP_001276470.1 | 9 | 2864 | |||
NM_001289542.1 | NP_001276471.1 | 9 | 3241 | |||
NM_011363.3 | NP_035493.2 | 10 | 3341 | |||
GenBank mRNA | AF020526.1 | - | 10 | 2982 | ||
AF074329.1 | - | 9 | 2686 | |||
AF380422.1 | - | 9 | 2935 | |||
AF421138.1 | - | 9 | 2882 | |||
AF421139.1 | - | 10 | 3035 | |||
AI182992.1 | - | 1 | 45 | |||
AK148085.1 | - | 11 | 3015 | |||
AK154655.1 | - | 10 | 3222 | |||
AK168439.1 | - | 10 | 2883 | |||
AK170444.1 | - | 10 | 3315 | |||
AK173145.1 | - | 5 | 962 | |||
AK195282.1 | - | 1 | 307 | |||
AK200463.1 | - | 1 | 303 | |||
BC011422.1 | - | 10 | 2833 | 3156603 | ||
BC051978.1 | - | 9 | 2784 | 6401205 |