Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ski | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011385.2 | NP_035515.2 | 1 | 947 | ||
GenBank mRNA | AF435852.1 | - | 1 | 926 | ||
BC068305.1 | - | 2 | 791 | 30093626 | ||
CA451127.1 | - | 1 | 271 | |||
CA750434.1 | - | 1 | 588 |