Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011397.4 | NP_035527.3 | 4 | 496 | ||
GenBank mRNA | AF058318.1 | - | 4 | 406 | ||
AK085499.1 | - | 4 | 411 | |||
AK143959.1 | - | 4 | 497 | |||
AK165439.1 | - | 4 | 432 | |||
BC013528.1 | - | 4 | 425 | 4238222 |