Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011402.3 | NP_035532.2 | 2, 3 | 154 | ||
GenBank mRNA | AF081499.1 | - | 2, 3 | 151 | ||
AK004832.1 | - | 2, 3 | 153 | |||
AK145599.1 | - | 2, 3 | 153 | |||
AK164786.1 | - | 2, 3 | 155 | |||
BB873139.1 | - | 2, 3 | 155 | |||
BC096369.1 | - | 1, 2 | 125 | 3962784 |