Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011426.3 | NP_035556.3 | 2 | 963 | ||
GenBank mRNA | BC141335.1 | - | 2 | 557 | 9055947 | |
BC141336.1 | - | 2 | 553 | 9055948 | ||
Z36233.1 | - | 2 | 961 | |||
Z36234.1 | - | 2 | 961 | |||
Z36293.1 | - | 2 | 961 |