Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109995.1 | NP_001103465.1 | 8 | 676 | ||
NM_011431.3 | NP_035561.1 | 8 | 673 | |||
GenBank mRNA | AK052242.1 | - | 8 | 631 | ||
AK150158.1 | - | 8 | 645 | |||
AK153473.1 | - | 8 | 645 | |||
AK154878.1 | - | 8 | 655 | |||
AK165624.1 | - | 8 | 647 | |||
AK172875.1 | - | 8 | 627 | |||
AK180021.1 | - | 1 | 27 | |||
AK184658.1 | - | 1 | 38 | |||
AY512947.1 | - | 1 | 1744 | |||
BC052674.1 | - | 8 | 649 | 30061580 | ||
BC054778.1 | - | 8 | 639 | 6810127 | ||
U97079.1 | - | 8 | 615 |