Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011499.3 | NP_035629.2 | 2 | 426 | ||
GenBank mRNA | AF096285.1 | - | 3 | 238 | ||
AK075804.1 | - | 2 | 384 | |||
AK077504.1 | - | 2 | 378 | |||
AK145971.1 | - | 2 | 378 | |||
AK147179.1 | - | 2 | 427 | |||
AK152283.1 | - | 2 | 276 | |||
AK154158.1 | - | 2 | 394 | |||
AK207782.1 | - | 1 | 24 | |||
BC141076.1 | - | 2 | 377 | 9055688 | ||
BC145342.1 | - | 2 | 377 | 9053898 |