Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011517.2 | NP_035647.2 | 3 | 282 | ||
GenBank mRNA | AK131949.1 | - | 3 | 206 | ||
BC048483.1 | - | 3 | 205 | |||
BC089540.1 | - | 3 | 205 | |||
BC099552.1 | - | 3 | 205 | |||
BC132079.1 | - | 3 | 283 | 40130356 | ||
BC138509.1 | - | 3 | 283 | 8861529 | ||
Y08485.1 | - | 3 | 291 |