Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ski | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011385.2 | NP_035515.2 | 2 | 1067 | ||
GenBank mRNA | AF435852.1 | - | 2 | 1046 | ||
AK019148.2 | - | 2 | 124 | |||
BC068305.1 | - | 3 | 911 | 30093626 | ||
CA451127.1 | - | 2 | 391 |