Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177561.1 | NP_001171032.1 | 2 | 573 | ||
NM_011387.2 | NP_035517.1 | 2 | 573 | |||
GenBank mRNA | AB003303.1 | - | 2 | 498 | ||
BC021154.1 | - | 2 | 535 | 4162098 | ||
BC094023.1 | - | 3 | 646 | 4160598 | ||
U95131.1 | - | 2 | 478 | |||
U95132.1 | - | 2 | 478 |