Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005443.4 | NP_005434.4 | 4 | 520 | ||
XM_011532400.1 | XP_011530702.1 | 4 | 500 | |||
XM_011532401.1 | XP_011530703.1 | 4 | 586 | |||
GenBank mRNA | AF016496.1 | - | 4 | 436 | ||
AF033026.1 | - | 4 | 462 | |||
AF105227.1 | - | 4 | 490 | |||
AK292774.1 | - | 4 | 470 | |||
AU132232.1 | - | 4 | 520 | |||
BC011392.1 | - | 4 | 393 | 3869484 | ||
BC032513.1 | - | 4 | 489 | |||
BC040940.1 | - | 4 | 448 | |||
BC050627.1 | - | 4 | 456 | 5556165 | ||
BQ437924.1 | - | 4 | 448 | |||
CR457028.1 | - | 4 | 436 | |||
Y10387.1 | - | 4 | 472 |