Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011394.3 | NP_035524.2 | 11 | 2928 | ||
GenBank mRNA | AK087507.1 | - | 11 | 2753 | ||
AK190097.1 | - | 1 | 168 | |||
AK193353.1 | - | 1 | 260 | |||
AK193654.1 | - | 1 | 169 | |||
BC046510.1 | - | 11 | 2848 | 4035182 | ||
BC094562.1 | - | 11 | 3308 |