Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011397.4 | NP_035527.3 | 7 | 845 | ||
GenBank mRNA | AF058318.1 | - | 7 | 755 | ||
AK085499.1 | - | 7 | 760 | |||
AK143959.1 | - | 7 | 846 | |||
AK165439.1 | - | 7 | 781 | |||
BC013528.1 | - | 7 | 774 | 4238222 |