Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011401.4 | NP_035531.3 | 5 | 1014 | ||
GenBank mRNA | AK045328.1 | - | 5 | 790 | ||
AK147258.1 | - | 4 | 3326 | |||
AK155997.1 | - | 5 | 773 | |||
AK164286.1 | - | 5 | 786 | |||
BC034122.1 | - | 5 | 772 | 4527667 | ||
BC050095.1 | - | 5 | 768 | |||
BC058811.1 | - | 5 | 772 | 6490844 | ||
CT010354.1 | - | 4 | 659 | |||
M75135.1 | - | 5 | 738 | |||
X61093.1 | - | 5 | 738 | |||
X69698.1 | - | 203 |