Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161853.1 | NP_001155325.1 | 2 | 406 | ||
NM_011418.2 | NP_035548.1 | 2 | 406 | |||
GenBank mRNA | AB017343.1 | - | 2 | 187 | ||
AB017344.1 | - | 2 | 187 | |||
AB041578.1 | - | 2 | 361 | |||
AJ011739.1 | - | 2 | 380 | |||
AJ011740.1 | - | 2 | 380 | |||
AK075906.1 | - | 2 | 389 | |||
AK145552.1 | - | 2 | 395 | |||
AK150169.1 | - | 2 | 407 | |||
BC025163.1 | - | 2 | 360 | 3154501 |