Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172472.1 | NP_001165943.1 | 3 | 582 | ||
NM_001172473.1 | NP_001165944.1 | 3 | 468 | |||
NM_001172475.1 | NP_001165946.1 | 3 | 507 | |||
NM_011451.3 | NP_035581.1 | 1 | 239 | |||
NM_025367.6 | NP_079643.2 | 3 | 465 | |||
GenBank mRNA | AF068748.1 | - | 3 | 507 | ||
AF068749.1 | - | 1 | 239 | |||
AF415213.1 | - | 1 | 137 | |||
AK028667.1 | - | 1 | 2529 | |||
AK136289.1 | - | 2 | 253 | |||
AK155332.1 | - | 3 | 567 | |||
AK160900.1 | - | 3 | 458 | |||
AK197304.1 | - | 2 | 152 | |||
BC011432.1 | - | 1 | 440 | |||
BC037710.1 | - | 3 | 380 | 4221357 |