Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011508.1 | NP_035638.1 | 2 | 299 | ||
GenBank mRNA | AF129888.1 | - | 2 | 284 | ||
AK148032.1 | - | 2 | 296 | |||
AK151070.1 | - | 2 | 296 | |||
AK151575.1 | - | 2 | 298 | |||
AK152194.1 | - | 2 | 298 | |||
AK169763.1 | - | 2 | 297 | |||
BC003463.1 | - | 2 | 299 | 2648657 | ||
BC010791.1 | - | 2 | 277 | 4218816 | ||
BC081429.1 | - | 2 | 286 | 5710951 | ||
BC096656.1 | - | 2 | 274 | 5143414 | ||
BC096691.1 | - | 2 | 284 | 5256396 | ||
Z50159.1 | - | 1 | 42 |