Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011512.3 | NP_035642.1 | 5 | 609 | ||
GenBank mRNA | AK004691.2 | - | 5 | 524 | ||
AK128902.1 | - | 5 | 523 | |||
AK145670.1 | - | 5 | 550 | |||
AK150568.1 | - | 5 | 427 | |||
AK151161.1 | - | 5 | 479 | |||
AK151319.1 | - | 5 | 437 | |||
AK152692.1 | - | 5 | 519 | |||
AK187611.1 | - | 4 | 294 | |||
BC003280.1 | - | 5 | 508 | 3582114 | ||
BC027352.1 | - | 5 | 469 | 3594564 | ||
M63114.1 | - | 5 | 1025 |