Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000744.6 | NP_000735.1 | 6 | 2003 | ||
NM_001256573.1 | NP_001243502.1 | 6 | 1934 | |||
NR_046317.1 | 6 | 2028 | ||||
XM_011528524.1 | XP_011526826.1 | 5 | 1647 | |||
XM_017027624.1 | XP_016883113.1 | 7 | 2035 | |||
XM_017027625.1 | XP_016883114.1 | 7 | 2053 | |||
GenBank mRNA | AK296430.1 | - | 2 | 274 | ||
AK314907.1 | - | 7 | 1719 | |||
BC096290.1 | - | 6 | 1911 | 40007296 | ||
BC096291.1 | - | 7 | 1819 | 40007297 | ||
BC096292.1 | - | 6 | 1842 | 40007298 | ||
BC096293.3 | - | 6 | 1936 | 40007299 | ||
DA530118.1 | - | 2 | 300 | |||
L35901.1 | - | 6 | 1950 | |||
U62433.1 | - | 6 | 2003 | |||
X87629.1 | - | 5 | 1697 | |||
X89746.1 | - | 1 | 90 | |||
Y08421.1 | - | 6 | 1772 |