Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011387.2 | NP_035517.1 | 4 | 1085 | ||
GenBank mRNA | U95132.1 | - | 4 | 990 |