Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011390.2 | NP_035520.1 | 18 | 2478 | ||
GenBank mRNA | AF087436.1 | - | 18 | 2480 | ||
AK131129.1 | - | 18 | 2432 | |||
AK143535.1 | - | 18 | 2472 | |||
AK149750.1 | - | 18 | 2473 | |||
AK164405.1 | - | 1 | 972 | |||
AK166215.1 | - | 18 | 2472 | |||
AK169950.1 | - | 18 | 2468 | |||
AK171498.1 | - | 18 | 2515 | |||
BC059242.1 | - | 18 | 2470 | |||
BC060133.1 | - | 18 | 2470 | |||
BC079677.1 | - | 15 | 4228 | |||
BC141107.1 | - | 18 | 2438 | 9055719 |