Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166450.2 | NP_001159922.1 | 11 | 1439 | ||
NM_001290703.1 | NP_001277632.1 | 13 | 1628 | |||
NM_001290704.1 | NP_001277633.1 | 13 | 1619 | |||
NM_001290705.1 | NP_001277634.1 | 10 | 1194 | |||
NM_011398.3 | NP_035528.1 | 11 | 1526 | |||
GenBank mRNA | AF076981.1 | - | 11 | 1527 | ||
AF535150.1 | - | 13 | 1620 | |||
AF535151.1 | - | 13 | 1629 | |||
AK148277.1 | - | 11 | 2974 | |||
BC048692.1 | - | 10 | 1249 | 6333237 |