Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011401.4 | NP_035531.3 | 10 | 2068 | ||
GenBank mRNA | AK028041.1 | - | 10 | 1844 | ||
AK045328.1 | - | 10 | 1844 | |||
AK147258.1 | - | 9 | 4380 | |||
AK155997.1 | - | 10 | 1827 | |||
AK164286.1 | - | 10 | 1840 | |||
BC034122.1 | - | 10 | 1826 | 4527667 | ||
BC050095.1 | - | 10 | 1822 |