Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109995.1 | NP_001103465.1 | 28 | 3265 | ||
NM_011431.3 | NP_035561.1 | 28 | 3262 | |||
GenBank mRNA | AK052242.1 | - | 28 | 3220 | ||
AK150158.1 | - | 28 | 3233 | |||
AK153473.1 | - | 28 | 3234 | |||
AK154878.1 | - | 28 | 3244 | |||
AK165624.1 | - | 28 | 3236 | |||
AK172875.1 | - | 28 | 3216 | |||
AK192797.1 | - | 1 | 353 | |||
AK194657.1 | - | 1 | 310 | |||
AK218919.1 | - | 1 | 317 | |||
BC012636.1 | - | 15 | 1977 | |||
BC036344.1 | - | 88 | ||||
BC052674.1 | - | 28 | 3238 | 30061580 | ||
BC054778.1 | - | 28 | 3228 | 6810127 | ||
U97079.1 | - | 28 | 3204 |