Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011454.1 | NP_035584.1 | 7 | 1000 | ||
GenBank mRNA | AK078753.1 | - | 7 | 1077 | ||
AK084785.1 | - | 7 | 1008 | |||
AK154048.1 | - | 7 | 1014 | |||
AK171741.1 | - | 6 | 837 | |||
BC061050.1 | - | 7 | 1007 | 30276988 | ||
U96707.2 | - | 7 | 1000 |