Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011499.3 | NP_035629.2 | 10 | 1712 | ||
GenBank mRNA | AF096285.1 | - | 11 | 1527 | ||
AK075804.1 | - | 10 | 1670 | |||
AK077504.1 | - | 10 | 1664 | |||
AK145971.1 | - | 10 | 1664 | |||
AK147179.1 | - | 10 | 1714 | |||
AK152283.1 | - | 10 | 1562 | |||
AK154158.1 | - | 10 | 1681 | |||
AK199765.1 | - | 1 | 329 |