Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011500.2 | NP_035630.2 | 15 | 1989 | ||
GenBank mRNA | AJ223777.1 | - | 15 | 1916 | ||
AK135684.1 | - | 10 | 1182 | |||
AK136032.1 | - | 13 | 1638 | |||
BC090968.1 | - | 15 | 1989 | 30646517 | ||
BC150727.1 | - | 15 | 1957 | 9087638 |