Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161722.1 | NP_001155194.1 | 2 | 394 | ||
NM_001161723.1 | NP_001155195.1 | 2 | 388 | |||
NM_011549.3 | NP_035679.3 | 1 | 335 | |||
GenBank mRNA | AF079095.1 | - | 1 | 316 | ||
AK078755.1 | - | 2 | 326 | |||
AK146016.1 | - | 1 | 360 | |||
AK154971.1 | - | 2 | 392 | |||
BC064789.1 | - | 1 | 335 | 30429545 |