Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alyref | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011568.1 | NP_035698.1 | 3 | 515 | ||
GenBank mRNA | AJ252140.1 | - | 3 | 212 | ||
BC120588.1 | - | 3 | 607 | 40129511 | ||
BC137658.1 | - | 3 | 607 | 8860677 | ||
U89876.1 | - | 3 | 515 |