Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011581.3 | NP_035711.2 | 3 | 467 | ||
GenBank mRNA | AK132508.1 | - | 3 | 298 | ||
AK142552.1 | - | 3 | 441 | |||
BC053702.1 | - | 3 | 269 | 30062369 | ||
CJ049934.1 | - | 3 | 468 | |||
L07803.1 | - | 3 | 295 | |||
M64866.1 | - | 3 | 295 |