Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164078.1 | NP_001157550.1 | 3 | 422 | ||
NM_001164079.1 | NP_001157551.1 | 3 | 416 | |||
NM_011585.4 | NP_035715.1 | 3 | 416 | |||
GenBank mRNA | AK009502.1 | - | 2 | 89 | ||
AK009914.1 | - | 2 | 89 | |||
AK028340.1 | - | 2 | 89 | |||
AK088486.1 | - | 3 | 352 | |||
AK134724.1 | - | 2 | 2749 | |||
AK144222.1 | - | 3 | 369 | |||
BC023813.1 | - | 3 | 331 | 5353938 | ||
BC027826.1 | - | 3 | 258 | |||
BC046812.1 | - | 3 | 305 | 5702183 | ||
U00689.1 | - | 3 | 357 | |||
U55862.1 | - | 3 | 224 |