Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011592.2 | NP_035722.2 | 2 | 150 | ||
GenBank mRNA | AK153866.1 | - | 2 | 154 | ||
BC023454.1 | - | 2 | 126 | |||
BC043479.1 | - | 2 | 130 | |||
BC110677.1 | - | 2 | 148 | 4979986 | ||
BC117523.1 | - | 2 | 140 | 40105563 | ||
BC117524.1 | - | 2 | 140 | 40105561 | ||
CA493459.1 | - | 2 | 188 | |||
U69898.1 | - | 2 | 140 |