Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011602.5 | NP_035732.2 | 5 | 922 | ||
GenBank mRNA | AK040128.1 | - | 2 | 138 | ||
AK122420.2 | - | 5 | 548 | |||
AK147228.1 | - | 5 | 493 | |||
BC150810.1 | - | 5 | 593 | 9087721 | ||
X56123.1 | - | 5 | 594 |