Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000834.3 | NP_000825.2 | 2 | 340 | ||
XM_011520628.2 | XP_011518930.1 | 3 | 1133 | |||
XM_011520629.2 | XP_011518931.1 | 4 | 940 | |||
XM_017019219.1 | XP_016874708.1 | 3 | 688 | |||
GenBank mRNA | BC113618.1 | - | 2 | 239 | 8322670 | |
BC113620.1 | - | 2 | 239 | 8322672 | ||
U11287.1 | - | 2 | 371 | |||
U28861.1 | - | 1 | 161 | |||
U88963.1 | - | 2 | 340 | |||
U90278.1 | - | 2 | 370 |