Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277903.1 | NP_001264832.1 | 8 | 337 | ||
NM_001277904.1 | NP_001264833.1 | 7 | 301 | |||
NM_011618.2 | NP_035748.1 | 8 | 334 | |||
GenBank mRNA | AF020946.1 | - | 8 | 321 | ||
AJ131711.1 | - | 7 | 282 | |||
AK160226.1 | - | 8 | 335 | |||
BC138871.1 | - | 8 | 314 | 8861893 | ||
BC141142.1 | - | 8 | 317 | 9055754 | ||
BC145449.1 | - | 7 | 281 | 9054007 | ||
BC145450.1 | - | 7 | 281 | 9054008 | ||
BC145451.1 | - | 8 | 317 | 9054009 | ||
U92883.1 | - | 8 | 318 | |||
U92884.1 | - | 7 | 285 |