Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011638.4 | NP_035768.1 | 3 | 331 | ||
GenBank mRNA | AK048228.1 | - | 3 | 245 | ||
AK088961.1 | - | 3 | 245 | |||
BC054522.1 | - | 3 | 216 | 6406097 | ||
BY101284.1 | - | 3 | 332 | |||
M29618.1 | - | 1 | 97 | |||
X57349.1 | - | 2 | 167 |