Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011645.2 | NP_035775.1 | 3 | 330 | ||
GenBank mRNA | AK131862.1 | - | 3 | 331 | ||
BC098498.1 | - | 3 | 313 |