Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcte3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011560.3 | NP_035690.2 | 1, 2 | 266 | ||
NM_198104.2 | NP_932772.1 | 1, 2 | 266 | |||
GenBank mRNA | AK006370.1 | - | 131 | |||
AY172988.1 | - | 145 | ||||
BC048739.1 | - | 195 | ||||
BC061136.1 | - | 149 | 6774834 | |||
BC131981.1 | - | 1, 2 | 257 | 40130258 | ||
BC131983.1 | - | 1, 2 | 257 | 40130260 | ||
BU919334.1 | - | 32 | ||||
CN835332.1 | - | 261 | ||||
CN842925.1 | - | 227 | ||||
M26332.1 | - | 197 | ||||
U21673.1 | - | 237 | ||||
U21674.1 | - | 1, 2 | 222 |
Gene Symbol: Gm3417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123368.1 | NP_001116840.1 | 259 |
Gene Symbol: Gm3448 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123367.1 | NP_001116839.1 | 1, 2 | 259 |