Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098226.3 | NP_001091696.2 | 3, 4 | 345 | ||
NM_001204156.1 | NP_001191085.1 | 3, 4 | 668 | |||
NM_011566.4 | NP_035696.3 | 3, 4 | 509 | |||
GenBank mRNA | AF002670.1 | - | 3, 4 | 464 | ||
AK087649.1 | - | 3, 4 | 346 | |||
AK143551.1 | - | 3, 4 | 367 | |||
BC021364.1 | - | 3, 4 | 389 | |||
BC047929.1 | - | 3, 4 | 668 | |||
CF617964.1 | - | 3, 4 | 509 | |||
D87964.1 | - | 3, 4 | 490 | |||
Y10027.1 | - | 2, 3 | 271 | |||
Y16611.1 | - | 2, 3 | 262 |