Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044384.1 | NP_001037849.1 | 4 | 389 | ||
NM_001294280.2 | NP_001281209.1 | 3 | 309 | |||
NM_011593.2 | NP_035723.2 | 4 | 386 | |||
GenBank mRNA | AA184230.1 | - | 2 | 155 | ||
BC008107.1 | - | 4 | 291 | 3158222 | ||
BC034260.1 | - | 4 | 291 | 3158222 | ||
BC051260.1 | - | 4 | 271 | 30056565 | ||
CJ055052.1 | - | 4 | 301 | |||
CJ063120.1 | - | 4 | 387 | |||
M17243.1 | - | 4 | 252 | |||
X04684.1 | - | 4 | 328 |