Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011631.1 | NP_035761.1 | 2 | 225 | ||
GenBank mRNA | AK146885.1 | - | 2 | 291 | ||
AK150811.1 | - | 2 | 245 | |||
AK151413.1 | - | 2 | 242 | |||
AK153550.1 | - | 2 | 245 | |||
AK159974.1 | - | 2 | 250 | |||
AK160093.1 | - | 2 | 245 | |||
AK165975.1 | - | 2 | 245 | |||
AK168952.1 | - | 2 | 245 | |||
AK169090.1 | - | 2 | 245 | |||
BC010445.1 | - | 2 | 157 | 3587178 | ||
BC011439.1 | - | 2 | 231 | 3707817 | ||
J03297.1 | - | 2 | 225 | |||
M16370.1 | - | 2 | 183 |