Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286122.1 | NP_001273051.1 | 2 | 367 | ||
NM_011632.3 | NP_035762.2 | 3 | 478 | |||
GenBank mRNA | AK147304.1 | - | 2 | 295 | ||
AK147362.1 | - | 2 | 368 | |||
BC137634.1 | - | 2 | 193 | 8860653 | ||
BC137635.1 | - | 2 | 193 | 8860654 | ||
CJ127244.1 | - | 3 | 386 | |||
U21050.1 | - | 3 | 297 |