Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011645.2 | NP_035775.1 | 4 | 541 | ||
GenBank mRNA | AK131862.1 | - | 4 | 542 | ||
BC098498.1 | - | 4 | 524 | |||
BC141304.1 | - | 4 | 606 | 9055916 |
Gene Symbol: Gm10334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103153.1 | NP_001096623.1 | 4 | 557 |
Gene Symbol: Gm5771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038997.2 | NP_001034086.1 | 4 | 555 | ||
GenBank mRNA | BC132026.1 | - | 4 | 555 | 40130303 | |
BC132030.1 | - | 4 | 555 | 40130307 |