Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164071.1 | NP_001157543.1 | 3 | 372 | ||
NM_001164072.1 | NP_001157544.1 | 2 | 167 | |||
NM_011529.2 | NP_035659.1 | 2 | 163 | |||
GenBank mRNA | AK031559.1 | - | 2 | 129 | ||
AK038910.1 | - | 2 | 102 | |||
AK088817.1 | - | 2 | 113 | |||
AK147727.1 | - | 3 | 318 | |||
AK165420.1 | - | 2 | 123 | |||
AK165994.1 | - | 2 | 98 | |||
BB660164.1 | - | 2 | 168 | |||
BC026148.1 | - | 2 | 148 | 3492937 | ||
BC033282.1 | - | 2 | 99 | |||
BC052685.1 | - | 2 | 150 | |||
U51907.1 | - | 2 | 105 | |||
U59864.1 | - | 2 | 176 |