Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285472.1 | NP_001272401.1 | 6 | 875 | ||
NM_001285476.1 | NP_001272405.1 | 4 | 799 | |||
NM_011532.2 | NP_035662.1 | 4 | 803 | |||
GenBank mRNA | AF326960.2 | - | 4 | 803 | ||
AF349658.1 | - | 6 | 875 | |||
BC139474.1 | - | 4 | 803 | 8862496 | ||
BC145636.1 | - | 4 | 799 | |||
U15565.1 | - | 2 | 200 | |||
U57327.1 | - | 4 | 425 |