Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285498.1 | NP_001272427.1 | 4 | 465 | ||
NM_001285500.1 | NP_001272429.1 | 4 | 318 | |||
NM_011565.2 | NP_035695.1 | 4 | 423 | |||
GenBank mRNA | AK132009.1 | - | 3 | 358 | ||
AK137064.1 | - | 3 | 1706 | |||
AK143155.1 | - | 3 | 1639 | |||
AK144857.1 | - | 4 | 385 | |||
AK146595.1 | - | 4 | 385 | |||
BC050217.1 | - | 3 | 243 | 30021393 | ||
D50563.1 | - | 4 | 423 | |||
X94442.1 | - | 3 | 337 | |||
Y10026.1 | - | 4 | 387 |